J.T.Baker® BAKERBOND Speedisk® Extraction Disks and Stations
Disks are preassembled for use in preparing aqueous samples for analysis.
Laminar configuration provides filtration capacity and inlet characteristics that maximize access of analyte molecules to the micro-particulate sorbent. Disk design resists clogging and ensures high throughput rates even when samples contain solids.
Frequently Bought Together