Lensx® 90 Lens Tissues, Berkshire
Fournisseur: Berkshire
Certificats
À propos de cet article
Lensx® Lens Tissues are very soft, non-abrasive tissues for use on most sensitive surfaces, such as lenses.
- Low in ash and organic impurities
- Tissues are polywrapped
Ideal for coated optics and laser cleaning applications. Nonwoven wipers are made of 70% rayon and 30% cellulose.
Certifications: Tissues comply with Government Lens Paper Specification AA-50177, Type 1, Lightweight ISO Class 7 (FED-STD-209E Class 10,000/M5.5) cleanroom applications.
Frequently Bought Together