Sand Viper Protective Eyewear, U.S. Safety
Supplier: Dentec Safety Specialists
Certificates
About this item
The Sand Viper's unique design permits quick and easy conversion from glasses to goggles, providing users with the means to adapt to any environment requiring protective eyewear.
- For rugged use providing protection, comfort, and a secure fit
- Frame features closed-cell memory foam technology with interior sculpting
- Perforated molding regulates air entry during rigorous activity
- PC lenses offer 99.9% UV protection
- Antifog, abrasion-resistant coating
Blue mirror lenses reduce glare and reflect heat, and are recommended for outdoor use where excessive UV levels are present. Clear lenses provide maximum visibility and protect against harmful UV levels. Gray lenses protect the user from glare and high levels of hazardous visible light and UV radiation without distorting color perception.
Available with +1.5 and +2.0 magnifying powers in clear lens styles.
Certifications: ANSI Z87.1
Ordering information: All glasses are supplied with a nylon pouch and headstrap for conversion purposes.
Frequently Bought Together