Citation 932 Series Safety Glasses, U.S. Safety
Supplier: Dentec Safety Specialists
Certificates
About this item
The Citation series of protective eyewear combines a modern, stylish appearance with all-day comfort and rugged durability.
- Polycarbonate lenses with a scratch-resistant coating
- Lenses provide 99.9% UV protection
These safety glasses features twin spherical lenses integrated into a single-lens design, molded nosepiece, and ratchet-adjustable temples.
Certifications: Meet ANSI Z87.1 and CSA Z94.3 specifications.
Frequently Bought Together