KleenGuard™ 30-06 Dual Readers Safety Glasses, Kimberly-Clark Professional
Supplier: Ansell Healthcare
Certificates
About this item
Trust KleenGuard™ dual readers safety glasses for an ideal combination of vision assistance and eye protection. They feature integrated diopters on both the top and the bottom for vision help when looking both up and down.
- Black frames with clear lenses
- Integrated diopters ('cheaters') on the top and bottom of the lenses, for vision assistance when looking up and down
- Safety readers have a 7 base curve, which facilitates peripheral vision
- Lenses are hardcoated to prevent damage and provide 99.9% UVA/UVB/UVC protection
- Ergonomic frame and comfortable
The stylish, black nylon frames feature a soft, self-adjusting rubber nose pad for comfort. The clear lenses allow maximum visible light transmission (ideal for indoor use) and are hardcoated for scratch resistance and longer life.
This design helps to prevent neckstrain, eyestrain, and headaches for individuals who work above their heads or upside down.
Certifications: Meets ANSI / ISEA Z87.1+ standard for personal eye protection. TAA compliant.
Frequently Bought Together