OPTI-MAX® Check Valves and Replacement Cartridges, Optimize Technologies
About this item
Preassembled check valve cartridges conveniently insert into a reusable housing and function in either the inlet or outlet position
Conversion kits include the necessary check valves and cartridges to convert a system with OEM style check valves to the OPTI-MAX® cartridge check valve system.
The two-piece Free-Turn™ design allows the outside housing thread to spin independently of the tube fitting port so the housing can be removed from the pump head with the tubing attached. A single cartridge is interchangeable across multiple pump brands. Ceramic ball and seat sets are precision-matched to exacting standards.
9 Options Available Below
Product Details & Documents
Preassembled check valve cartridges conveniently insert into a reusable housing and function in either the inlet or outlet position
Conversion kits include the necessary check valves and cartridges to convert a system with OEM style check valves to the OPTI-MAX® cartridge check valve system.
The two-piece Free-Turn™ design allows the outside housing thread to spin independently of the tube fitting port so the housing can be removed from the pump head with the tubing attached. A single cartridge is interchangeable across multiple pump brands. Ceramic ball and seat sets are precision-matched to exacting standards.
Product Family Options
Product Information
- DésignationMatièreFor Use WithAvailabilityPrice
- Active Inlet Valve (AIV) Replacement Cartridge, 600 barStainless steel, PEEK, ZirconiaHP/Agilent 1050 /1100 /1200Log in for availabilityEach (1Inch)$887.12Supplier Number: 27-38-04713
- Supplier Number: 27-38-00672
- Supplier Number: 27-38-00680
- HP/Agilent 1090Discontinued-Supplier Number: -
- Supplier Number: 27-38-01959
- Supplier Number: 27-38-00682
- Outlet Cartridge Check Valve, 1/8" Ceramic Ball and SeatStainless steelHP/Agilent 1050, 1100, 1200, 1220, 1260Log in for availabilityEach$1,025.30Supplier Number: 27-56-01330
- Outlet Cartridge Check Valve, 1/8" Ceramic Ball and SeatPEEKHP/Agilent 1050, 1100, 1200, 1220, 1260Log in for availabilityEach$1,023.30Supplier Number: 27-58-01330
- Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire SeatStainless steelHP/Agilent 1050, 1100, 1200, 1220, 1260Log in for availabilityEach$1,020.77Supplier Number: 27-57-01330
- Outlet Cartridge Check Valve, 1/8" Ruby Ball/Sapphire SeatPEEKHP/Agilent 1050, 1100, 1200, 1220, 1260Log in for availabilityEach$1,020.77Supplier Number: 27-59-01330