Pierce Qentix™ Western Blot Signal Enhancer, Thermo Scientific
Catalog # CAPI21050
Supplier: Thermo Scientific
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Western Blot Signal Enhancer
- Size:500 mL
- Cat. no.:CAPI21050
- Supplier No.:21050
Specifications
About this item
Western blot signal enhancer is used to pre-treat either nitrocellulose or PVDF membranes for a 3 – 10X increase in signal intensity for most target proteins.
- Protein targets can be detected at lower levels
- Enhances antibody binding to immobilized antigens
- Works with any substrate, enhancing both chemiluminescence and colorimetric detection
- Works with any membrane
- Ready-to-use and can be stored at room temperature
Signal enhancement has been demonstrated with targets such as IL-6, p53, NF[kappa]B, BRCA1, and EGF.