Rubicon™ Pro Grade Protective Eyewear, MCR Safety
Fournisseur: MCR Safety
Certificats
À propos de cet article
These glasses feature a curved, 180° single lens design with a close orbital seal for increased clarity and protection.
- Lenses are scratch-resistant and filter out 99% of UV radiation
- Platinum color metal temples with resilient spring hinges provide maximum flexibility
- Soft, secure gel nosepiece and nonslip gel temple sleeves provide a comfortable fit for extended wear
- Antifog lenses feature exclusive Duramass AF4® antifog lens coating
Certifications: Eyewear meets or exceeds ANSI Z87+ standards.
Ordering information: Each pair comes with a breakaway cord and a black satin carrying case that can be used as a cleaning cloth.
Frequently Bought Together