To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Chemiluminescent Nucleic Acid Detection Module
- Cat. no.:CAPI89880
- Supplier No.:89880
- Sufficient For:800 cm2 of membrane (10 Mini-Gel Blots)
Specifications
About this item
A complete system for detection of biotin-labeled nucleic acids for various blotting applications including Northern/Southern blots, ribonuclease protection assays (RPAs), electrophoretic mobility shift assays (EMSAs or gel shifts).
- Fast and sensitive
- Versatile kit protocol (included) is applicable to any biotin-based nucleic acid detection system: Northerns, Southerns, EMSA, RPAs and colony lifts
- Kit contains only the critical detection components for those who already have biotin-labeled probes and an optimized hybridization method developed
It is designed for those researchers who already have biotin-labeled probes and an optimized hybridization method developed to allow for quick and sensitive detection results in a wide variety of protocol applications.
The system uses an enhanced luminol substrate for horseradish peroxidase (HRP) with optimized blocking and wash steps that together produce sensitivity equivalent to radioactive (32P) systems and provide a clean, consistent background required for publication-quality data.