BD BBL™ Isolator Pack Plated Media, BD Diagnostics
BD BBL™ Isolator Pack Plated Media, BD Diagnostics
 :   BD
BD BBL™ Isolator Pack Plated Media, BD Diagnostics
 :  BD
BD BBL™ Isolator Pack Plated Media, BD Diagnostics
BD BBL™ Isolator Pack Plated Media, BD Diagnostics
BD BBL™ prepared agar plates in isolator packs are plated media in gamma-irradiated packs that are specially designed for isolator systems
  • Specially designed for isolator systems
  • Validated for performance and sterility according to the Association for the Advancement of Medical Instrumentation (AAMI) guidelines
To use, place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle. When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping. Use the included inner bag to transport plates from your critical environment. Storage temperature: 2 to 8 °C.
The multi-wrap packaging protects media from vaporized hydrogen peroxide exposure.
Ordering information: Each pack or case contains a signed, lot-specific Certificate of Analysis.
  • Catalog No:
  • CA-90004-234
  • CA-90007-914
  • CA-90007-920
  • Sterility:
  • Sterile
  • Sterile
  • Sterile
Product Family Options
  • Désignation
    Plate Diameter
    Type de conditionnement
  • R2A Agar
    100 mm
    Isolator pack, triple wrapping, VHP resistant
    Pack of 100
    Pack of 100 - $1,340.00
    $1,340.00
    Pack of 100
       $1,340.00
    Minimum order quantity is 1
     : 292341
    Specifications
    Cat. No.
    CA-90004-234
    Sterility
    Sterile
  • Trypticase™ Soy Agar with Lecithin and Polysorbate 80, Finger Dab
    100 mm
    Isolator pack, triple wrapping, VHP resistant
     : -
    Specifications
    Cat. No.
    CA-90007-914
    Sterility
    Sterile
  • Isolator Pack Trypticase™ Soy Agar
    100 mm
    Isolator pack, triple wrapping, VHP resistant
    Pack of 100
    Pack of 100 - $751.00
    $751.00
    Pack of 100
       $751.00
    Minimum order quantity is 1
     : 292652
    Specifications
    Cat. No.
    CA-90007-920
    Sterility
    Sterile