PEEK Sample Loops, Rheodyne®
Fournisseur: Rheodyne
Certificats
À propos de cet article
These loops are designed for Rheodyne models 9125, 9725, and 9010 PEEK injectors.
Loops are supplied with fitting that are unswaged so that the tube may be completely bottomed in the injector port before the ferrule is swaged on. Accuracy of large loops (0.030" bore) is about 14%, intermediate loops (0.020") 21%, and small loops (0.007") 65%. The size designations are nominal, and the actual volumes can differ from the nominal designations due to the ±0.002" tolerance of the tubing bore.
Frequently Bought Together