Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Fundamentals of physics infinite photogate adapter
- Cat. no.:470352-192
Extend your photogate to accommodate any width.
- Clips securely to the Fundamentals of Physics Photogate system
- Two mirrored adapters allow both photogates to be set up with extra long gate lengths
- Perfect for a wide variety of physics experiments, such as free fall, uniform circular motion, and attwood machines
Want to analzye the motion of an object that is too large to fit through the arms of a photogate? Whether it's a tennis ball in free fall or a student-constructed sail cart, this simple device allows you to create a photogate with an 'infinite' path length. The mirrored clips attach easily and securely to the infrared 'source' side of the photogate. A red laser pointer can then be aimed at the mirror so that the beam reflects into the detector side of the photogate. If an object blocks the laser's beam, the photogate will register an 'event' as usual.