ContactUsLinkComponent
Wall Bracket for 32 oz. Bottles, R & R Lotion
  56614-158
Small Business Enterprise
undefined
Wall Bracket for 32 oz. Bottles, R & R Lotion
  56614-158
 :  WALL-BR-32
Small Business Enterprise
  • Description:
    Wall Bracket for 946 ml (32 oz.) bottles
  • Cat. no.:
    56614-158
  • Supplier No.:
    WALL-BR-32
Steel wall brackets are designed for use with any of R & R Lotion's 946 mL (32 oz.) products
This includes lotions, sunscreens, hand cleaners, moisturizing hand sanitizers, and barrier lotions.