UVP Blak-Ray® UV Protective Spectacles, Analytik Jena
Catalog # 33002-067
Supplier: Analytik Jena US
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Blak-Ray® UV protective spectacles, 51H×152W mm
- Cat. no.:33002-067
- Supplier No.:98-0002-01
Specifications
About this item
Specially formulated plastic lenses block transmission of 254, 302, and 365 nm UV light to virtually zero.
The 'blue haze' normally associated with longer wavelength ultraviolet light is eliminated. Special sideshields help protect against peripheral exposure, making Blak-Ray® spectacles ideal for observing demonstrations or for visitor protection.