Norton 180® Protective Eyewear, Honeywell Safety
Catalog # CA89032-860
Supplier: Honeywell Safety Products
Specifications
- Cat. no.:CA89032-860
- Lens Tint:Clear
- Supplier No.:T18000
Specifications
About this item
Wraparound styling combines maximum protection with on-the-job comfort.
- Eyewear is 100% lightweight polycarbonate with a clear
- Scratch-resistant
- One-piece wraparound lens
Vented side shield temples reduce heat buildup and lens fogging. Frosted brow reduces glare from overhead lighting. Four-post hinge system increases impact resistance by dispersing impact among additional contact points.
Certifications: ANSI Z87.1 / CSA Z94.3