Solvent Filter/Degasser, MilliporeSigma | Avantor
Solvent Filter/Degasser, MilliporeSigma
Supplier:   MilliporeSigma
Solvent Filter/Degasser, MilliporeSigma
Supplier:  MilliporeSigma
Certificates
Solvent Filter/Degasser, MilliporeSigma
Solvent Filter/Degasser, MilliporeSigma
1 Options Available Below
Product Details & Documents
Used for HPLC.
Attaches directly to an empty, one-gallon solvent bottle. Designed to filter particulates that could damage expensive pumps and detectors and to remove dissolved oxygen in the solvent that could cause detector drift and instability. Filters and degasses one gallon of solvent in 10 minutes without operator attention.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • HPLC Solvent Filter/Degasser
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,063.74
    $6,063.74
    Each
       $6,063.74
    Supplier Number: 692000-1
  • Replacement Filter, PTFE Membrane
    Discontinued
    Supplier Number: -