Synthware Flask, Pear-Shaped, Threaded, Microscale, Kemtech America
Supplier: Kemtech America
Synthware Flask, Pear-Shaped, Threaded, Microscale, Kemtech America
Supplier: Kemtech America
Certificates
Jump to:
À propos de cet article
Pear-shaped threaded flask for microscale applications.
- Heavy wall borosilicate construction
- Threaded joint
- Compression cap included
Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.
3 Options Available Below
Product Details & Documents
Pear-shaped threaded flask for microscale applications.
- Heavy wall borosilicate construction
- Threaded joint
- Compression cap included
Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.
Product Family Options
Informations sur le produit
Accessories
Informations sur le produit
- Catalog #MaterialCap SizeAvailabilityPrice
- Nylon cap
Informations sur le produit
- Catalog #I.D.×WO-Ring SizeAvailabilityPrice
- Viton® O-Ring, 14/10 threaded joints