Synthware Hickman Distillation Head, with Side Port, Microscale, Kemtech America
60004-138
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Cat. no.:60004-138
- ST Joint Size:14/10
- Supplier No.:H121410
Hickman still head with side port for syringe access.
- Heavy wall borosilicate construction
- Threaded upper joint
- Open cap included
- Hickman Syle still head with side port
For microscale fractional distillation. Supplied with a nylon compression cap, Viton* O-ring, and side port cap with 8 mm PTFE-lined silicone septum.