Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America | Avantor
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
14/10
Catalog # 60004-114
Supplier:   Kemtech America
Fournisseur #:  A621410
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
Catalog # 60004-114
Supplier:  Kemtech America
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
Synthware Adapter, Vacuum Takeoff, Threaded, Microscale, Kemtech America
14/10
Price
$94.11
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
This is a vacuum takeoff joint adapter suitable for microscale applications.
  • Threaded
  • Nylon compression cap and Viton® O-ring included
Delivery information: Supplied with a nylon compression cap and Viton® O-ring.
Spécifications
  • ST Joint:
    14/10
Accessories
Informations sur le produit
  • Catalog #
    Material
    Cap Size
    Availability
    Price
  • Nylon cap
    Nylon cap
    60005-054
    Nylon
    20 mm
    Log in pour la disponibilité
    Each - 4,65 $
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring, 14/10 threaded joints
    Viton® O-Ring, 14/10 threaded joints
    60004-998
    014
    Log in pour la disponibilité
    Each - 5,61 $