Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America | Avantor
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
14/10
Catalog # 60004-106
Supplier:   Kemtech America
Fournisseur #:  A781410
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
Catalog # 60004-106
Supplier:  Kemtech America
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
Synthware Adapter, Thermometer, with Hose Connection, Microscale, Kemtech America
14/10
Price
$71.71
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Vacuum joint adapter with vertically adjustable condenser tube for microscale applications.
  • Vacuum-tight seal
  • Cap accommodates thermometer
  • Nylon compression cap and Viton® O-ring included
  • With hose connection
Supplied with a nylon compression cap and Viton® O-ring to form a vacuum tight seal. To use as a syringe port, replace the O-ring with PTFE -lined septum. Cap accommodates a 5.0 to 7.5 mm (ext. Ø) thermometer.
Spécifications
  • ST Joint:
    14/10
Accessories
Informations sur le produit
  • Catalog #
    Material
    Cap Size
    Availability
    Price
  • Nylon cap
    Nylon cap
    60005-052
    Nylon
    13 mm
    Log in pour la disponibilité
    Each - $2.73
Informations sur le produit
  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring
    Viton® O-Ring
    60004-994
    6.07×1.78 mm
    010
    Log in pour la disponibilité
    Each - $2.67