Synthware Adapter, Inlet or Thermometer, Microscale, Kemtech America
60004-104
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- ST Joint:14/10
- Environmentally Preferable:
- Cat. no.:60004-104
- Supplier No.:A771410
This is a joint adapter suitable for microscale applications.
- Vacuum-tight seal
- Cap accommodates thermometer
- Nylon compression cap and Viton® O-ring included
Supplied with a nylon compression cap and Viton® O-ring to form a vacuum tight seal. To use as a syringe port, replace the O-ring with PTFE -lined septum. Cap accommodates a 5.0 to 7.5 mm ext. Ø thermometer.