Ultra IBD 3 µm and 5 µm Columns, Restek
: Restek
Ultra IBD columns assist in deactivating surface silanols, and help to provide separation selectivities for acids, bases, zwitterions, and other polar compounds.
- Unique selectivity
- High level of base deactivation
- Without endcap
Intrinsically base-deactivated (IBD) phase columns contain a polar group within, or intrinsic to, the hydrocarbon-bonded phase. This results in unique selectivity and a high level of base deactivation, while reducing or eliminating the need for mobile phase additives.
- Catalog No:
- 89047-786
- 89047-798
- 89047-794
- 89047-790
- 89047-834
- 89047-810
- 89047-858
- 89047-850
- 89047-854
- 89047-802
- 89047-806
- 89047-822
- 89047-814
- 89047-862
- 89047-818
- 89047-826
- 89047-842
- 89047-838
- 89047-830
- 89047-846
- Description:
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Ultra IBD Columns
- Carbon load:
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- pH range:
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- 2.5 to 7.5
- Temperature limit:
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
- 80°C (176°F)
Frequently Bought Together