Formaldehyde Solidifier Spill Kit, NPS
Spill Kit
  46000-162
 :   NPS
 :  481001
Formaldehyde Solidifier Spill Kit, NPS
  46000-162
 :  NPS
Formaldehyde Solidifier Spill Kit, NPS
Formaldehyde Solidifier Spill Kit, NPS
Spill Kit
$273.76
Each
Spill kit is designed to provide a safe method of cleaning up small formaldehyde leaks or spills
Spill kit comes with one 0.91kg (2lbs.) shaker bottle of formaldehyde solidifier, one 0.91kg (2lbs.) shaker bottle of AQUALOCKIT® solidifier for pH neutral liquids, two 30.5x30.5cm (12x12") hazmat pads, one scoop with detachable scraper, one pair nitrile gloves, one pair goggles, one disposal bag with twist tie, and one disposal label.
Note: Recommended for use by trained personnel only.
Ideal for a quick response in spill-prone areas. Solidifier suppresses the vapor while solidifying the formaldehyde for easier disposal.
  • Désignation:
    Spill Kit