Safety Cleaning Wipes, Alcohol Free, Bausch & Lomb®
47745-036
- Description:Safety Cleaning Wipes
- Cat. no.:47745-036
- Supplier No.:8595
Cleaning wipes work on all safety equipment and are safe for use on rubberized materials.
They are premoistened with benzalkonium chloride and come in a dispenser that holds 100 individually packaged 12.7×20.3 cm (5×8") wipes. Wipes aid employees in meeting OSHA safety equipment care guidelines.