À propos de cet article
Sonifier® Cell Disrupters are designed for biological and liquid processing applications where precise control and parameter measurement are required.
- Adjustable amplitude
- LCD display
- Separate stop and pause buttons
- Five different language setup menus
Front panel features an 80-character LCD display of parameter settings, alarms, and messages, and a bargraph display of relative power while the unit is running
Analog units feature a 0–15 minute timer with "Hold" position, duty cycle control with pulser mode for applying pulses of energy at one pulse a second (pulse duration variable from 0.1 to 0.9 seconds) and a continuous "On" position. Meter indicates percentage of ultrasonic power at any output setting.Soundproof enclosure reduces noise level up to 20db when liquids are treated ultrasonically. Especially helpful when using a cell disrupter for extended periods.
Preset up to 20 different parameter setups. Separate front panel stop and pause buttons for experimental cycles. Select from five different language setup menus (English, German, Italian, French, or Spanish). Parallel printer interface and serial interface for RS-232. Printed report capability with date stamp and parameter information. Self-diagnostics check performed on power-up.
Spécifications
- Catalog No:
- CA33995-320
- CA33995-319
- CA33995-594
- CA33995-312
- CA33995-309
- CA33995-454
- CA33995-584
- CA33995-310
- CA33995-588
- CA33995-592
- Model:
- S-450A
- S-450A
- S-250D
- S-450A
- S-250A
- S-450D
- S-450D
- S-450A
- S-450D
- S-250D
- Volume range:
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL
- 1–500 mL