Accessories for SFX550 Sonifier® Cell Disruptor, with 1/2" Horn, Branson
CA33996-469
- Description:Soundproof Enclosure
- Cat. no.:CA33996-469
- Interior Dimensions:32.4W x 26.7D x 52.7H cm (123/4 x 101/2 x 203/4")
- Exterior Dimensions:39.4W x 39.4D x 59.7H cm (151/2 x 151/2 x 231/2")
- Weight:22.7 kg (50 lbs.)
- Supplier No.:101-063-275