ContactUsLinkComponent
Lens Cleaning Towelettes, C-Clear
  89012-762
 :  C-Clear
undefined
Lens Cleaning Towelettes, C-Clear
  89012-762
 :  C-Clear
 :  70BP
  • Description:
    Bulk-Packed Towelettes
  • Cat. no.:
    89012-762
  • Packaging:
    1000/Box
  • Supplier No.:
    70BP
Cardboard dispenser holds 100 individually wrapped 12.7×20.3 cm (5×8") towelettes that are treated with anti-fog/anti-static formula.
  • Sturdy corrugated cardboard dispenser
  • Pull-out bottom for easy dispensing
  • Built-in tab for hanging
  • Maintain eyewear properly with these premoistened towelettes
  • Ideal for cleaning glass, plastic, and polycarbonate lenses
May also be used on side shields, face shields, goggles, respirators, CRT screens, and microscopes.
 : Bulk packed towelettes are supplied without cardboard dispensers.