To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:VersaHOCl™ sterile disinfectant and cleaner, Double bagged
- Cat. no.:77530-434
- Packaging:Trigger spray bottle
- Volume:946 ml (6.32 oz.)
Specifications
About this item
VersaHOCl™ Sterile is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire’s VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
- Kills common microbiological contaminations on surfaces
- Can be used in a variety of applications
- Sterile filled and provided double bagged for easy transfer into critical environments
- Product certificate provided with each case
- Ideal for large surface cleaning
- No strong odor provides good user acceptability
- No special disposal is required
- Facilitates transfer disinfection into cleanroom
- No mixing required
Non-toxic with low odor and low residue, VersaHOCl™ establishes the safest standards in disinfectant technology. With an active ingredient of 0.065% hypochlorous acid, it’s non-corrosive and effective on hard, non-porous surfaces. This versatile disinfectant is ideal for use in controlled environments such as labs, cleanrooms, compounding pharmacies, and manufacturing facilities. It can also be used in areas such as hospitals, clinics, healthcare facilities, schools, and government and office buildings.