KIMBLE® KIMAX® Pink Colorware Erlenmeyer Flasks, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Please join us to support breast cancer prevention, treatment, and research by purchasing Pink Colorware from DWK Life Sciences.
- Manufactured from borosilicate glass 3.3
- With durable ceramic enamel markings to indicate approximate volumes at various levels
- Pink markings are brighter in appearance than traditional glassware with white markings
- Tops are reinforced and tooled with a rounded finish
- Body is thick-walled, with a long tapered outside contour to minimize chipping when struck or rubbed together
Choose from a wide variety of KIMBLE® KIMAX® Pink “Colorware”, including Low Form Griffin Beakers, Narrow Mouth Erlenmeyer Flasks, Class A Volumetric Flasks, Blood Typing Tubes, and GL45 caps for ease of identification in the lab.
We will donate 20% of the proceeds to the Breast Cancer Research Foundation, the highest rated breast cancer organization in the United States. BCRF is recognized as one of the most financially efficient nonprofits in the country, and we are proud to assist in their support of breast cancer research.
Complies with ASTM Specification E1404, Type I, Class I requirements.
Frequently Bought Together