ContactUsLinkComponent
KIMBLE® KIMAX® Pink Colorware Beakers, Low Form, DWK Life Sciences
  10017-910
 : 14000P-400
undefined
KIMBLE® KIMAX® Pink Colorware Beakers, Low Form, DWK Life Sciences
  10017-910
 :  14000P-400
 : 14000P-400
Select an Option
77 mm
Select an Option
25 ml
Select an Option
50 - 325 ml
Select an Option
400 ml
Select an Option
Pink Low Form Griffin Beakers
Select an Option
106 mm
Select an Option
10017-910
Select an Option
14000P-400
Please join us to support breast cancer prevention, treatment, and research by purchasing Pink Colorware from DWK Life Sciences.
  • With spout
  • Manufactured from borosilicate glass 3.3
  • Use of colored beakers reduces the risk of cross contamination
  • Easily identify glassware between labs
  • All sizes feature double capacity scales with easy-to-read graduations
  • Durable, color, matte finish marking area for use with an ordinary pencil or marker
Choose from a wide variety of KIMBLE® KIMAX® Pink “Colorware”, including Low Form Griffin Beakers, Narrow Mouth Erlenmeyer Flasks, Class A Volumetric Flasks, Blood Typing Tubes, and GL45 caps for ease of identification in the lab.
We will donate 20% of the proceeds to the Breast Cancer Research Foundation, the highest rated breast cancer organization in the United States. BCRF is recognized as one of the most financially efficient nonprofits in the country, and we are proud to assist in their support of breast cancer research.
Complies with ASTM Specification E960, Type I requirements.
Frequently Bought Together