Compresstome® Starter Kits | Avantor
Compresstome® Starter Kits
Compresstome® Starter Kits
Certificates
Compresstome® Starter Kits
Compresstome® Starter Kits
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Compresstome starter kit or VF-800-0Z starter kit.
  • Starter kits for Compresstome vibrating microtome or model VF-800-0Z
  • Allows for simple set-up
  • Includes everything needed to complete tissue slicing
The Compresstome vibrating microtome starter kit includes ten stainless steel blades, ten agarose tablets, one packet petroleum jelly, two plastic tissue pipettes, one super glue, one machine oil (for maintenance), two fine tissue forceps, and five plastic clean-up razors.
The Compresstome VF-800-0Z starter kit includes two tungsten carbide blades, 100 agarose tablets, two super glue, two fine tissue forceps, five plastic clean-up razors, three fine tissue handling brushes, and ten plastic tissue pipettes.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Compresstome starter kit for VF-800-0Z
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $964.87
    $964.87
    Each
       $964.87
    Supplier Number: VF8000ZSKUSA
  • Compresstome starter kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $390.58
    $390.58
    Each
       $390.58
    Supplier Number: VFSKVMCANAL