Accessories for Compresstome® Buffer Tray | Avantor
Accessories for Compresstome® Buffer Tray
Accessories for Compresstome® Buffer Tray
Certificates
Accessories for Compresstome® Buffer Tray
Accessories for Compresstome® Buffer Tray
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel Compresstome buffer tray is typically used for sterile sectioning procedures.
  • Buffer tray is autoclavable
  • Full one year warranty
Replacement gasket and screws set for the buffer tray for Compresstome tissue slicers. This set includes one set of screws and one gasket.
Compresstome buffer tray made of high-quality, durable stainless steel. Autoclavable. Full one year warranty. This set includes, one stainless steel buffer tray, one set of screws, one gasket, and one small Allen wrench.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Compresstome buffer tray gasket and screws
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $33.48
    $33.48
    Each
       $33.48
    Supplier Number: VFBTGAS
  • Compresstome stainless steel buffer tray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $839.85
    $839.85
    Each
       $839.85
    Supplier Number: VFBTVMSSBOS