About this item
Compresstome Specimen Tube Kits are compatible with the Compresstome VF-510-0Z.
- Specimen tube kits are available in 15.5 or 20 mm diameters
Compresstome specimen tube kits with 15.5 or 20 mm diameter include two specimen tubes with matching plungers, one chilling block, and one buffer tray.
Compresstome specimen tube kits with 15 or 20 mm diameter accessories and a blade holder include two specimen tubes with matching plungers, one chilling block, one buffer tray, and one blade holder.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.