ContactUsLinkComponent
Compresstome® Magnetic Blade Holder
  77435-416
undefined
Compresstome® Magnetic Blade Holder
  77435-416
 :  VFBHVMBOS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Compresstome VF-210-0Z and VF-510-0Z blade holder
  • Material:
    Stainless steel, magnetic
  • Cat. no.:
    77435-416
Replacement blade holder for Compresstome vibrating microtome.
  • Each blade holder securely holds ceramic and/or steel blades
  • Blade holder attaches to vibrating head, and is easily removable
For use on the VF-210-0Z or VF-510-0Z Compresstome vibratome, the magnetic blade holder features magnets for easy attachment of stainless steel blades. It is compatable with ceramic blades as well.
 : Includes 1 blade holder, 1 Allen wrench, and 1 small screw.