Compresstome® Blades
À propos de cet article
Compresstome stainless steel or ceramic blades.
- All blades are compatible with the compresstome models
Stainless steel blades are double-edged. They are packaged 5 double-edged blades per box (two boxes per order).
Ceramic blades for cutting tissue with compresstome slicers. Ceramic blades are durable and long-lasting, typically good for up to 3 to 4 weeks based on 1 sample per day. Sold as a single blade.
2 Options Available Below
Product Details & Documents
Compresstome stainless steel or ceramic blades.
- All blades are compatible with the compresstome models
Stainless steel blades are double-edged. They are packaged 5 double-edged blades per box (two boxes per order).
Ceramic blades for cutting tissue with compresstome slicers. Ceramic blades are durable and long-lasting, typically good for up to 3 to 4 weeks based on 1 sample per day. Sold as a single blade.