Accessories for Compresstome® Oxygenation Attachment
Catalogue # 77435-530
Fournisseur: PRECISIONARY INSTRUMENTS LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Oxygenation attachment
- Cat. no.:77435-530
Spécifications
À propos de cet article
This is an oxygenation tube compatible with compresstome models, used to assist in the process of keeping samples alive longer.
- Clips onto the buffer tank to help buffer solutions stay oxygenated during the cutting process
- Used for experiments involving fresh/acute/live tissue slices that require oxygen
Oxygenation attachment used to attach to the buffer tray of the compresstome tissue slicers.