Accessories for Compresstome® Legacy Buffer Tray Plug
Product Details & Documents
The compresstome legacy buffer tray plug is compatible with compresstome models VF-200, VF-200-0Z, VF-300, and VF-300-0Z.
- Buffer tray plug to prevent solution drainage
- Full one year warranty
This buffer tray plug is used to prevent the drainage of buffer solution from the buffer tray.
Spécifications
- Désignation:Compresstome legacy buffer tray plug