Accessories for Compresstome® Buffer Tray Kits for VF-310-0Z and VF-210-0Z
Fournisseur: PRECISIONARY INSTRUMENTS LLC
Certificats
À propos de cet article
Buffer tray kits for use with compresstome models VF-310-0Z and VF-210-0Z.
- Buffer trays are made from high quality, durable Lexan™ polycarbonate
- Available tube diameter sizes: 15.5 or 20 mm
- Full one year warranty
Compatable with compresstome models VF-310-0Z or VF-210-0Z, these 15.5 or 20 mm.
Ordering information: Include one buffer tray, one set of screws, and one gasket.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together