Accessories for Rotary Microtome Plastic Casettes and Metal Molds
Fournisseur: PRECISIONARY INSTRUMENTS LLC
Certificats
À propos de cet article
Plastic casettes and metal molds for tissue sample fixation and preparation.
- Rotary microtome components for tissue sample fixation and preparation
- Casettes are used for embedding fixed tissue settings in wax prior to tissue sectioning
- Molds are used with plastic embedding casettes for embedding samples in wax
The casette is supplied with bottom and lid.
Metal molds are highly durable and reusable while allowing efficient heat/cold exchange. The molds will accommodate large and small tissue samples, including a wide variety of tissue thicknesses.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together