Compresstome® Sectioning Tools
Compresstome® Sectioning Tools
Compresstome® Sectioning Tools
Compresstome® Sectioning Tools
Sectioning tools designed for use with compresstome rotary microtomes.
  • Fine tissue forceps are supplied individually
  • Tissue paintbrushes are supplied in packs of 3
Fine tissue handling forceps enable gentle manipulation of tissues and organs during dissection. They allow for the gentle removal of the tissue from the bufffer tray while slicing.
Tissue paintbrushes allow for easy handling of delicate tissue slices. Soft bristles help prevent damage to tissue sections when transferring slices.
Product Family Options
  • Désignation
  • Fine tissue handling forceps
    Each
    Each - $28.17
    $28.17
    Each
       $28.17
     : VFFORCEPS
  • Fine tissue handling brushes
    Pack of 3
    Pack of 3 - $118.63
    $118.63
    Pack of 3
       $118.63
     : VFVMPBCANAL