Compresstome® VF-510-0Z, VF-300-0Z, or VF-210-0Z vibratomes.
- Compresstome® VF-510-0Z is fully automated to cut slices in 1 µm increments and includes a 5 year warranty
- Compresstome® VF-300-0Z is fully automated for sectioning fixed and live tissues and includes a 5 year warranty
- Compresstome® VF-210-0Z is a vibrating microtome for cutting tissue slices with Auto Zero-Z technology
The VF-510-0Z is the newest Compresstome® model and is fully automated. This vibratome comes calibrated with Auto Zero-Z patented technology, which ensures healthier live slices and smoother fixed sections. This model comes with a one-touch unlocking mechanism on the sliding control box that allows for rapid sample loading and cutting.
The VF-300-0Z is a fully automated Compresstome® vibratome. It comes with a one year warranty and does not have the Auto-Zero Z technology.
The VF-210-0Z is a vibrating microtome for cutting tissue slices with Auto Zero-Z technology.
The VF-300-0Z includes VF-300 vibrating microtome, one removable blade holder, and one specimen tube kit.
The VF-210-0Z model includes VF-210-0Z vibrating microtome, one removable magnetic blade holder, and one specimen tube kit.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.