Compresstome® Gooseneck Lamp and Acessories
Supplier: PRECISIONARY INSTRUMENTS LLC
Certificates
About this item
The gooseneck lamp adds a spotlight when slicing specimens. The steel puck is used to attach the gooseneck lamp to the Compresstome® base.
- LED spot illuminator lamp with flexible gooseneck
The Compresstome® lamp includes one LED spotlight on a flexible gooseneck that allows for easy adjustment. The steel puck allows for the magnet base of the Gooseneck Lamp to adhere when inserted on the Compresstome® vibrating microtome models.
Packaging: One set of screws/washers for mounting included. Steel puck with attached bolt includes one nut for securing bolt.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together