ContactUsLinkComponent
Accessories for Rotary Microtome RF-1000 Side Panel
Catalog # 77435-310
undefined
Accessories for Rotary Microtome RF-1000 Side Panel
Catalog # 77435-310
Supplier Number:  RF1000SIDEBOX
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Rotary microtome RF-1000 side panel
  • Cat. no.:
    77435-310
Specifications
About this item
A replacement part for the external control panel on the RF-1000 rotary microtome.
  • Includes side control panel and cable
Control panel allows for control of slice thickness adjustments, movement of the specimen head, and start/stop of automated cutting.