Compresstome® Agarose tablets
Supplier: PRECISIONARY INSTRUMENTS LLC
Jump to:
À propos de cet article
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
- Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
4 Options Available Below
Product Details & Documents
Agarose tablets, each 0.5 mg, for easy use in specimen preparations for Compresstome® tissue samples.
- Tablets are available in packs of 10, 20, 50, or 100
Agarose is used for fresh or fixed samples to section using a Compresstome® rotary microtome.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: VFAGTVM10CANAL
- Supplier Number: VFAGTVM20CANAL
- Supplier Number: VFAGTVM50CANAL
- Supplier Number: VFAGT100CANAL