ContactUsLinkComponent
Micro Brite™ Pro LED Lighted Pocket Microscope with Adapter Clip
Catalog # 470355-712
Supplier:  CARSON OPTICAL
Micro Brite™ Pro LED Lighted Pocket Microscope with Adapter Clip
Catalog # 470355-712
Supplier:  CARSON OPTICAL
Supplier Number:  MM-350
Specifications
  • Magnification:
    60 - 250×
  • Model:
    MM-350
  • Description:
    Zoom LED pocket microscope, 2.3×1.2×3.7"
  • Weight:
    0.13 lbs (excluding battery)
  • Cat. no.:
    470355-712
  • Includes:
    Smartphone adapter clip
Specifications
About this item
This pocket microscope is perfect for exploring the world around you, with a 60 to 120× high magnification zoom and bright LED light to help you see all the small details. The high quality design of all Carson microscopes ensures this portable microscope both highly effective and durable. Crafted with precision molded aspheric lenses, the MicroBrite™ always provides premium optics with super bright, clear vision, guaranteeing the most accurate view of your magnified image. Whether you’re a professional scientist or just curious about the microscopic world around you, the Carson MicroBrite Pro portable microscope is the perfect tool for your next adventure!
  • Digiscoping
  • Lightweight compact design
  • Multi-Use
  • Used by any age
The MicroBrite™ Pro is also easy to use. Unlike traditional microscopes, the MicroBrite™ Pro does not require microscope slides to view objects. Begin by simply placing the object you wish to view directly below the objective lens of your pocket microscope. Next, turn on the LED light with the built in switch and adjust the upper zoom dial to find your desired magnification. Finally, rotate the lower Focus Ring until the image of your object is clear and sharp. The MicroBrite™ Pro features a 10 to 14mm exit pupil diameter to provide 60 to 120× magnification and 2× optical zoom. And thanks to the Smartphone Adapter, you may then easily clip your smartphone to the eyepiece of your MicroBrite™ Pro to utilize its digiscoping abilities and take digital photos or videos to share with your loved ones. The MicroBrite is battery powered with one AA battery.
With its durable yet lightweight design and high magnification, the Carson MicroBrite™ Pro is the best microscope for kids, adults, students of any age, teachers, scientists, or anyone who loves to explore the world around them. Whether you’re investigating the wonders of the natural world, getting a closer look at objects around the house, or performing professional science experiments, the MicroBrite™ Pro is up to the task. The Carson MicroBrite™ ensures quality in any application such as a STEM microscope for kids or a pocket microscope for coins, food, fabric, trichomes, or anything else you can find! Or use the MicroBrite in a professional setting for the appraisal of jewelry, currency or stamps, or observations in biology, chemistry, or geography. or a pocket microscope for coins, food, fabric, trichomes, or anything else you can find! Or use the MicroBrite™ in a professional setting for the appraisal of jewelry, currency or stamps, or observations in biology, chemistry, or geography.
Ordering information: Powered by 1 AA battery (not included).