Needle Cleaning Wire
Cleaning wire, 180 mm long and 0.13 mm Ø, Gauges G22s, G25, G28, G33
Catalog # 77506-630
Supplier: INNOVATIVE LABOR SYSTEME GMBH
Fournisseur #: 21018301
Jump to:
Product Details & Documents
Cleaning wire for gauges G22s, G25, G28, G33.
Spécifications
- Désignation:Cleaning wire, 180 mm long and 0.13 mm Ø
- For Use With:Gauges G22s, G25, G28, G33