Synthware Vacuum Traps with Spherical Joints
Supplier: Kemtech America
Jump to:
À propos de cet article
Traps feature 35/20 spherical joints for added flexibility.
- Heavy Wall Borosilicate Construction
- 35/20 Spherical Joint
- Adapters sold Separately
The trap body has a 50 mm ext. Ø and is 360 mm in length. Each of the three adapters features a 16 mm ext. Ø outlet. The three adapters and 35/20 stainless steel pinch clamps must be ordered separately.
4 Options Available Below
Product Details & Documents
Traps feature 35/20 spherical joints for added flexibility.
- Heavy Wall Borosilicate Construction
- 35/20 Spherical Joint
- Adapters sold Separately
The trap body has a 50 mm ext. Ø and is 360 mm in length. Each of the three adapters features a 16 mm ext. Ø outlet. The three adapters and 35/20 stainless steel pinch clamps must be ordered separately.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: V175039
- Supplier Number: V175038
- Supplier Number: V175037
- Supplier Number: V175036
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 2 mm PTFE Plug
Informations sur le produit
- Catalog #AvailabilityPrice
- PTFE plug, 4 mm
Informations sur le produit
- Catalog #AvailabilityPrice
- Clamps for 35/20 spherical joints