Synthware Single Bank Vacuum Manifold with #15 O-Ring Joints, Kemtech America
60004-594
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Description:Single bank vacuum manifold
- Cat. no.:60004-594
- No. of Ports:4
- Supplier No.:M311508
Manifold is equipped with four ports that are spaced 100 mm from center to center.
- Hi vacuum valves
- Heavy wall borosilicate construction
- Four ports
- #15 O-ring joints
The 0 to 8 mm high vacuum valves are mounted on a 45° angle to the operator for easy access. Overall width is 395 mm.
: Accessory items must be ordered separately.