Order Entry
Canada
ContactUsLinkComponent
Synthware Manifold with #15 O-Ring Joints, Kemtech America
Synthware Manifold with #15 O-Ring Joints, Kemtech America
Catalog # 60004-592
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Manifold with #15 O-Ring Joints, Kemtech America
Catalog # 60004-592
Supplier:  Kemtech America
Supplier Number:  M150004
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Description:
    Vacuum/Gas Manifold
  • Cat. no.:
    60004-592
  • No. of Ports:
    4
  • Supplier No.:
    M150004

Specifications

About this item

Manifold features 4 mm double hollow high vacuum stopcocks

  • Hollow glass stopcocks (held in by vacuum)
  • Heavy wall borosilicate construction
  • 4 Ports
  • Two barbed ports
  • Two ground glass points

The four-port manifold is fitted with two 14/20 and two 24/40 inner joints, with a distance of 100 mm from joint center to center.

Ordering information: All accessories must be ordered separately.