Synthware Connecting Adapters, Kemtech America
60000-322
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- ST Joint:24/40
- Environmentally Preferable:
- Cat. no.:60000-322
- Supplier No.:A192424
Converts standard vessels to airless applications.
- [ST] joint
- PTFE stopcock
Features a 2 mm PTFE stopcock.